The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257328.1 | NP_001244257.1 | 11 - 12 | 1207 | 67 |
NM_001257330.1 | NP_001244259.1 | 11 - 12 | 1144 | 67 | |
NM_001257331.1 | NP_001244260.1 | 10 - 11 | 1099 | 67 | |
NM_004313.3 | NP_004304.1 | 11 - 12 | 1144 | 67 | |
NM_199004.1 | NP_945355.1 | 10 - 11 | 1099 | 67 | |
NR_047516.1 | 10 - 11 | 1113 | 67 | ||
XM_006721520.1 | XP_006721583.1 | 9 - 10 | 930 | 67 | |
XM_011523858.2 | XP_011522160.1 | 11 - 12 | 1029 | 67 | |
XM_017024645.1 | XP_016880134.1 | 10 - 11 | 1016 | 67 | |
GenBank mRNA | AB209786.1 | - | 10 - 11 | 919 | 67 |
AF106941.1 | - | 11 - 12 | 969 | 67 | |
AK097542.1 | - | 10 - 11 | 959 | 67 | |
AK290951.1 | - | 10 - 11 | 900 | 67 | |
AK297059.1 | - | 10 - 11 | 977 | 67 | |
AK297181.1 | - | 11 - 12 | 1008 | 67 | |
BC007427.2 | - | 11 - 12 | 978 | 67 | |
BC039066.1 | - | 10 - 11 | 907 | 67 | |
BC067368.1 | - | 11 - 12 | 972 | 67 | |
CR450310.1 | - | 11 - 12 | 916 | 67 | |
CR749218.1 | - | 10 - 11 | 937 | 67 | |
DQ664180.1 | - | 11 - 12 | 916 | 67 | |
EU883572.1 | - | 11 - 12 | 916 | 67 | |
Z11501.1 | - | 11 - 12 | 916 | 67 |