The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030915.3 | NP_112177.2 | 1 - 2 | 236 | 74 |
GenBank mRNA | AK223541.1 | - | 1 - 2 | 172 | 74 |
AK314598.1 | - | 1 - 2 | 230 | 74 | |
AL136703.1 | - | 1 - 2 | 240 | 74 | |
BC109376.1 | - | 1 - 2 | 240 | 74 | |
BC126434.1 | - | 1 - 2 | 208 | 74 | |
BC130415.1 | - | 1 - 2 | 208 | 74 | |
EF025586.1 | - | 1 - 2 | 213 | 74 |