The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015909.3 | NP_056993.2 | 12 - 13 | 1126 | 80 |
NR_052013.2 | 12 - 13 | 1126 | 80 | ||
XM_011510358.2 | XP_011508660.1 | 12 - 13 | 1122 | 80 | |
XM_011510359.2 | XP_011508661.1 | 6 - 7 | 527 | 80 | |
XM_017004317.1 | XP_016859806.1 | 12 - 13 | 1122 | 80 | |
GenBank mRNA | AB621813.1 | - | 12 - 13 | 1126 | 80 |
AF388385.1 | - | 12 - 13 | 1108 | 80 | |
AL832774.1 | - | 12 - 13 | 1069 | 80 | |
AL833270.1 | - | 12 - 13 | 1087 | 80 | |
BC040345.1 | - | 4 - 5 | 394 | 80 | |
BC108693.1 | - | 12 - 13 | 1101 | 80 | |
BC131735.1 | - | 12 - 13 | 1108 | 80 |