Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBAS
Entrez Gene ID
51594
Gene Name
neuroblastoma amplified sequence
Gene Aliases
ILFS2, NAG, SOPH
Chromosome Location
Chr.2: 14998067 - 15561348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015909.3 NP_056993.2 28 - 29 3304 71
NR_052013.2 28 - 29 3304 71
XM_011510357.2 XP_011508659.1 27 - 28 3171 71
XM_011510358.2 XP_011508660.1 28 - 29 3300 71
XM_011510359.2 XP_011508661.1 22 - 23 2705 71
XM_011510360.2 XP_011508662.1 8 - 9 1124 71
XM_011510361.2 XP_011508663.1 8 - 9 1115 71
XM_017004317.1 XP_016859806.1 28 - 29 3300 71
GenBank mRNA AB621813.1 - 28 - 29 3304 71
AF056195.1 - 7 - 8 506 71
AF388385.1 - 28 - 29 3286 71
AL050281.1 - 1 - 2 73 71
AL833270.1 - 28 - 29 3264 71
BC040345.1 - 20 - 21 2572 71
BC051792.1 - 4 - 5 403 71
BC131735.1 - 28 - 29 3286 71

View Full Product Details