The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077199.2 | NP_001070667.1 | 6 - 7 | 1036 | 76 |
NM_001270492.1 | NP_001257421.1 | 7 - 8 | 1160 | 76 | |
NM_001323527.1 | NP_001310456.1 | 7 - 8 | 1160 | 76 | |
NM_001323529.1 | NP_001310458.1 | 6 - 7 | 1036 | 76 | |
NM_001323533.1 | NP_001310462.1 | 6 - 7 | 1036 | 76 | |
NM_001323534.1 | NP_001310463.1 | 7 - 8 | 1051 | 76 | |
NM_001323535.1 | NP_001310464.1 | 6 - 7 | 927 | 76 | |
NM_139168.3 | NP_631907.1 | 7 - 8 | 1189 | 76 | |
XM_011543171.2 | XP_011541473.1 | 6 - 7 | 1261 | 76 | |
XM_017009055.1 | XP_016864544.1 | 6 - 7 | 933 | 76 | |
XM_017009056.1 | XP_016864545.1 | 6 - 7 | 821 | 76 | |
GenBank mRNA | AF459094.1 | - | 6 - 7 | 870 | 76 |
AK091758.1 | - | 6 - 7 | 919 | 76 | |
AK308287.1 | - | 7 - 8 | 1063 | 76 | |
AL832888.1 | - | 3 - 4 | 1893 | 76 | |
BC047322.1 | - | 6 - 7 | 905 | 76 | |
BC067770.1 | - | 7 - 8 | 1190 | 76 | |
BC112343.1 | - | 4 - 5 | 558 | 76 |