The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012293.2 | NP_036425.1 | 9 - 10 | 1112 | 88 |
XM_005264707.3 | XP_005264764.1 | 8 - 9 | 1028 | 88 | |
XM_011510396.1 | XP_011508698.1 | 9 - 10 | 869 | 88 | |
XM_011510397.2 | XP_011508699.1 | 8 - 9 | 957 | 88 | |
GenBank mRNA | AF200348.1 | - | 9 - 10 | 1071 | 88 |
BC030834.1 | - | 9 - 10 | 1155 | 88 | |
BC098579.1 | - | 9 - 10 | 1007 | 88 | |
D86983.1 | - | 9 - 10 | 1071 | 88 | |
EF090903.1 | - | 9 - 10 | 1071 | 88 |