The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015356.4 | NP_056171.3 | 31 - 32 | 4339 | 127 |
NM_182706.4 | NP_874365.3 | 31 - 32 | 4339 | 127 | |
GenBank mRNA | AF240677.1 | - | 31 - 32 | 4332 | 127 |
AF271734.2 | - | 31 - 32 | 4310 | 127 | |
AY062238.1 | - | 31 - 32 | 4341 | 127 | |
BC009490.2 | - | 7 - 8 | 719 | 127 | |
BC014632.2 | - | 13 - 14 | 1938 | 127 | |
D63481.3 | - | 31 - 32 | 4334 | 127 |