The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006788.3 | NP_006779.1 | 5 - 6 | 1426 | 67 |
GenBank mRNA | AB209924.1 | - | 3 - 4 | 633 | 67 |
AY517554.1 | - | 3 - 4 | 692 | 67 | |
BC013126.1 | - | 5 - 6 | 1288 | 67 | |
L42542.1 | - | 5 - 6 | 1434 | 67 |