Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
Chromosome Location
Chr.17: 31094927 - 31377677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000267.3 NP_000258.1 15 - 16 2104 114
NM_001042492.2 NP_001035957.1 15 - 16 2104 114
XM_005257983.2 XP_005258040.1 15 - 16 2054 114
XM_005257984.2 XP_005258041.1 15 - 16 2054 114
XM_006721922.2 XP_006721985.1 16 - 17 2084 114
XM_006721924.2 XP_006721987.1 16 - 17 2084 114
XM_006721925.2 XP_006721988.1 16 - 17 2084 114
XM_006721926.3 XP_006721989.1 16 - 17 2084 114
XM_011524852.2 XP_011523154.1 16 - 17 2084 114
XM_011524857.2 XP_011523159.1 16 - 17 2084 114
XM_017024689.1 XP_016880178.1 16 - 17 2084 114
XM_017024690.1 XP_016880179.1 16 - 17 2084 114
GenBank mRNA BC144643.1 - 15 - 16 1823 114
BX537850.1 - 13 - 14 1496 114
M38106.1 - 7 - 8 719 114
M38107.1 - 7 - 8 719 114
M82814.1 - 15 - 16 1932 114
M89914.1 - 15 - 16 1918 114

View Full Product Details