The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 32 - 33 | 4748 | 129 |
NM_001042492.2 | NP_001035957.1 | 33 - 34 | 4811 | 129 | |
XM_005257983.2 | XP_005258040.1 | 33 - 34 | 4761 | 129 | |
XM_005257984.2 | XP_005258041.1 | 32 - 33 | 4698 | 129 | |
XM_006721922.2 | XP_006721985.1 | 34 - 35 | 4791 | 129 | |
XM_006721924.2 | XP_006721987.1 | 34 - 35 | 4791 | 129 | |
XM_006721925.2 | XP_006721988.1 | 33 - 34 | 4728 | 129 | |
XM_006721926.3 | XP_006721989.1 | 34 - 35 | 4791 | 129 | |
XM_011524852.2 | XP_011523154.1 | 34 - 35 | 4788 | 129 | |
XM_011524857.2 | XP_011523159.1 | 34 - 35 | 4791 | 129 | |
XM_017024689.1 | XP_016880178.1 | 33 - 34 | 4728 | 129 | |
GenBank mRNA | AB209336.1 | - | 15 - 16 | 2901 | 129 |
BC144643.1 | - | 32 - 33 | 4467 | 129 | |
M38106.1 | - | 24 - 25 | 3363 | 129 | |
M38107.1 | - | 24 - 25 | 3363 | 129 | |
M60915.1 | - | 7 - 8 | 927 | 129 | |
M61213.1 | - | 8 - 9 | 1098 | 129 | |
M82814.1 | - | 32 - 33 | 4576 | 129 | |
M89914.1 | - | 33 - 34 | 4625 | 129 |