Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
Chromosome Location
Chr.3: 52794768 - 52809009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002217.3 NP_002208.3 12 - 13 1606 73
XM_005265105.4 XP_005265162.1 12 - 13 1606 73
XM_017006356.1 XP_016861845.1 11 - 12 1512 73
GenBank mRNA AK222757.1 - 12 - 13 1578 73
AK223483.1 - 12 - 13 1604 73
BC107604.1 - 12 - 13 1576 73
BC107605.1 - 12 - 13 1576 73
BC107814.1 - 12 - 13 1576 73
X14690.1 - 4 - 5 538 73
X67055.1 - 12 - 13 1578 73

View Full Product Details