The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002217.3 | NP_002208.3 | 17 - 18 | 1977 | 77 |
XM_005265105.4 | XP_005265162.1 | 18 - 19 | 2088 | 77 | |
XM_017006356.1 | XP_016861845.1 | 16 - 17 | 1883 | 77 | |
GenBank mRNA | AK122858.1 | - | 1 - 2 | 1332 | 77 |
AK222757.1 | - | 17 - 18 | 1949 | 77 | |
AK223483.1 | - | 17 - 18 | 1975 | 77 | |
BC107604.1 | - | 17 - 18 | 1947 | 77 | |
BC107605.1 | - | 17 - 18 | 1947 | 77 | |
BC107814.1 | - | 17 - 18 | 1947 | 77 | |
X67055.1 | - | 17 - 18 | 1949 | 77 |