The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015527.3 | NP_056342.3 | 3 - 4 | 1184 | 104 |
XM_011545790.2 | XP_011544092.1 | 3 - 4 | 1241 | 104 | |
GenBank mRNA | AB449895.1 | - | 3 - 4 | 701 | 104 |
AF370370.1 | - | 3 - 4 | 329 | 104 | |
AK023192.1 | - | 3 - 4 | 563 | 104 | |
BC063112.1 | - | 3 - 4 | 608 | 104 | |
BC072453.1 | - | 3 - 4 | 496 | 104 | |
BC093814.1 | - | 3 - 4 | 412 | 104 | |
BC093816.1 | - | 3 - 4 | 412 | 104 | |
BC098419.1 | - | 3 - 4 | 748 | 104 |