Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D10B
Entrez Gene ID
26000
Gene Name
TBC1 domain family member 10B
Gene Aliases
EPI64B, FP2461
Chromosome Location
Chr.16: 30357101 - 30370258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015527.3 NP_056342.3 8 - 9 1817 71
XM_011545789.1 XP_011544091.1 5 - 6 1559 71
XM_011545790.2 XP_011544092.1 7 - 8 1684 71
GenBank mRNA AB449895.1 - 8 - 9 1334 71
AF370370.1 - 8 - 9 963 71
AK002101.1 - 2 - 3 320 71
AK023192.1 - 8 - 9 1196 71
AL117408.1 - 1 - 2 1014 71
BC050523.2 - 4 - 5 531 71
BC063112.1 - 8 - 9 1241 71
BC072453.1 - 8 - 9 1129 71
BC093814.1 - 8 - 9 1045 71
BC093816.1 - 8 - 9 1045 71
BC098419.1 - 8 - 9 1381 71

View Full Product Details