Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF3
Entrez Gene ID
23469
Gene Name
PHD finger protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 63635802 - 63715522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290259.1 NP_001277188.1 10 - 11 3661 73
NM_015153.3 NP_055968.1 8 - 9 3120 73
XM_005248701.3 XP_005248758.1 9 - 10 3455 73
XM_005248702.3 XP_005248759.1 8 - 9 3199 73
XM_005248703.3 XP_005248760.1 7 - 8 1409 73
XM_006715426.3 XP_006715489.1 9 - 10 3680 73
XM_011535648.2 XP_011533950.1 9 - 10 3337 73
XM_017010626.1 XP_016866115.1 8 - 9 1672 73
GenBank mRNA AF091622.1 - 8 - 9 3121 73
AK298580.1 - 7 - 8 1393 73
AK307869.1 - 7 - 8 3020 73
BC113650.1 - 8 - 9 3121 73
BC113652.1 - 8 - 9 3121 73
BX648268.1 - 10 - 11 3347 73
D87685.2 - 8 - 9 3109 73

View Full Product Details