The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113546.1 | NP_001107018.1 | 6 - 7 | 1044 | 92 |
NM_001113547.1 | NP_001107019.1 | 3 - 4 | 925 | 92 | |
NM_016357.4 | NP_057441.1 | 6 - 7 | 1044 | 92 | |
XM_011538455.2 | XP_011536757.1 | 6 - 7 | 1104 | 92 | |
GenBank mRNA | AB209512.1 | - | 6 - 7 | 980 | 92 |
AF198454.1 | - | 6 - 7 | 971 | 92 | |
AF198455.1 | - | 3 - 4 | 863 | 92 | |
AF218025.1 | - | 2 - 3 | 178 | 92 | |
AK000335.1 | - | 3 - 4 | 568 | 92 | |
AK000372.1 | - | 6 - 7 | 954 | 92 | |
AK222971.1 | - | 6 - 7 | 967 | 92 | |
AK314447.1 | - | 6 - 7 | 956 | 92 | |
AL136911.1 | - | 6 - 7 | 963 | 92 | |
BC001247.1 | - | 3 - 4 | 576 | 92 | |
BC110815.1 | - | 6 - 7 | 981 | 92 | |
BC136763.1 | - | 6 - 7 | 930 | 92 | |
BC144402.1 | - | 6 - 7 | 930 | 92 | |
BC144403.1 | - | 6 - 7 | 930 | 92 |