The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181435.5 | NP_852100.3 | 1 - 2 | 442 | 80 |
GenBank mRNA | AF173888.1 | - | 1 - 2 | 125 | 80 |
AK295968.1 | - | 1 - 2 | 442 | 80 | |
AK315921.1 | - | 1 - 2 | 429 | 80 | |
BC120990.2 | - | 1 - 2 | 372 | 80 | |
BX483406.1 | - | 1 - 2 | 389 | 80 | |
EU399232.1 | - | 1 - 2 | 306 | 80 |