The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006446.4 | NP_006437.3 | 9 - 10 | 1238 | 131 |
GenBank mRNA | AB026257.1 | - | 9 - 10 | 1233 | 131 |
AF060500.1 | - | 8 - 9 | 1134 | 131 | |
AF205071.1 | - | 9 - 10 | 1268 | 131 | |
AJ132573.1 | - | 8 - 9 | 1197 | 131 | |
AK312970.1 | - | 9 - 10 | 1220 | 131 | |
BC114376.1 | - | 9 - 10 | 1233 | 131 |