The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005966.3 | NP_005957.2 | 1 - 2 | 240 | 82 |
XM_017004173.1 | XP_016859662.1 | 1 - 2 | 125 | 82 | |
GenBank mRNA | BC035724.1 | - | 1 - 2 | 129 | 82 |