The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321315.1 | NP_001308244.1 | 2 - 3 | 975 | 115 |
XM_005246587.1 | XP_005246644.1 | 3 - 4 | 1008 | 115 | |
XM_017004175.1 | XP_016859664.1 | 3 - 4 | 1217 | 115 | |
GenBank mRNA | AF045452.1 | - | 2 - 3 | 975 | 115 |
AK296549.1 | - | 2 - 3 | 303 | 115 |