The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000395.2 | NP_000386.1 | 1 - 2 | 44 | 101 |
XM_005261340.3 | XP_005261397.1 | 1 - 2 | 103 | 101 | |
XM_011529903.2 | XP_011528205.1 | 1 - 2 | 103 | 101 | |
XM_011529904.2 | XP_011528206.1 | 1 - 2 | 103 | 101 | |
XM_011529905.2 | XP_011528207.1 | 1 - 2 | 103 | 101 | |
GenBank mRNA | BC070085.1 | - | 1 - 2 | 44 | 101 |