The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000395.2 | NP_000386.1 | 9 - 10 | 1370 | 86 |
XM_005261340.3 | XP_005261397.1 | 9 - 10 | 1447 | 86 | |
XM_011529903.2 | XP_011528205.1 | 9 - 10 | 1447 | 86 | |
XM_011529904.2 | XP_011528206.1 | 9 - 10 | 1429 | 86 | |
XM_011529905.2 | XP_011528207.1 | 9 - 10 | 1447 | 86 | |
GenBank mRNA | AK302993.1 | - | 7 - 8 | 1032 | 86 |
BC070085.1 | - | 9 - 10 | 1370 | 86 | |
CR456428.1 | - | 8 - 9 | 1244 | 86 | |
M59941.1 | - | 8 - 9 | 1181 | 86 |