Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL1A
Entrez Gene ID
114876
Gene Name
oxysterol binding protein like 1A
Gene Aliases
ORP-1, ORP1, OSBPL1B
Chromosome Location
Chr.18: 24162045 - 24397882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080597.3 NP_542164.2 5 - 6 607 117
XM_006722380.3 XP_006722443.1 4 - 5 520 117
XM_006722382.3 XP_006722445.1 5 - 6 620 117
XM_017025530.1 XP_016881019.1 5 - 6 899 117
XM_017025531.1 XP_016881020.1 4 - 5 392 117
XM_017025532.1 XP_016881021.1 4 - 5 392 117
GenBank mRNA AF323726.2 - 5 - 6 564 117
AF392450.1 - 5 - 6 564 117
AK096358.1 - 5 - 6 607 117
AK292895.1 - 5 - 6 539 117
BC136452.1 - 5 - 6 504 117
BC144117.1 - 5 - 6 504 117

View Full Product Details