Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HRG
Entrez Gene ID
3273
Gene Name
histidine rich glycoprotein
Gene Aliases
HPRG, HRGP, THPH11
Chromosome Location
Chr.3: 186665952 - 186678240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000412.4 NP_000403.1 6 - 7 817 85
GenBank mRNA AK313381.1 - 6 - 7 787 85
BC069574.1 - 6 - 7 760 85
BC150591.1 - 6 - 7 758 85
CR749302.1 - 4 - 5 3083 85
M13149.1 - 6 - 7 858 85

View Full Product Details