The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321193.1 | NP_001308122.1 | 15 - 16 | 1781 | 75 |
NM_001321194.1 | NP_001308123.1 | 16 - 17 | 1961 | 75 | |
NM_001321195.1 | NP_001308124.1 | 14 - 15 | 1678 | 75 | |
NM_024665.5 | NP_078941.2 | 15 - 16 | 1842 | 75 | |
XM_005247775.2 | XP_005247832.1 | 14 - 15 | 1797 | 75 | |
XM_006713746.1 | XP_006713809.1 | 16 - 17 | 1851 | 75 | |
XM_011513141.1 | XP_011511443.1 | 16 - 17 | 1785 | 75 | |
XM_011513142.2 | XP_011511444.1 | 15 - 16 | 1766 | 75 | |
XM_011513143.2 | XP_011511445.1 | 14 - 15 | 1692 | 75 | |
XM_017007185.1 | XP_016862674.1 | 15 - 16 | 1779 | 75 | |
GenBank mRNA | AF268193.1 | - | 15 - 16 | 1675 | 75 |
AF314544.1 | - | 14 - 15 | 1641 | 75 | |
AK022956.1 | - | 15 - 16 | 1741 | 75 | |
AK296946.1 | - | 13 - 14 | 1423 | 75 | |
BC113421.1 | - | 14 - 15 | 1584 | 75 | |
BX648080.1 | - | 1 - 2 | 1455 | 75 | |
BX648935.1 | - | 15 - 16 | 1776 | 75 |