The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321193.1 | NP_001308122.1 | 3 - 4 | 322 | 116 |
NM_001321194.1 | NP_001308123.1 | 4 - 5 | 502 | 116 | |
NM_024665.5 | NP_078941.2 | 3 - 4 | 383 | 116 | |
XM_005247775.2 | XP_005247832.1 | 2 - 3 | 338 | 116 | |
XM_006713746.1 | XP_006713809.1 | 4 - 5 | 392 | 116 | |
XM_011513141.1 | XP_011511443.1 | 4 - 5 | 326 | 116 | |
XM_011513142.2 | XP_011511444.1 | 3 - 4 | 307 | 116 | |
XM_011513143.2 | XP_011511445.1 | 2 - 3 | 233 | 116 | |
XM_017007185.1 | XP_016862674.1 | 3 - 4 | 320 | 116 | |
GenBank mRNA | AF268193.1 | - | 3 - 4 | 216 | 116 |
AF314544.1 | - | 2 - 3 | 182 | 116 | |
AK022956.1 | - | 3 - 4 | 282 | 116 | |
BC060320.1 | - | 2 - 3 | 236 | 116 | |
BC113421.1 | - | 2 - 3 | 125 | 116 | |
BX648935.1 | - | 3 - 4 | 317 | 116 | |
DB150466.1 | - | 4 - 5 | 401 | 116 | |
DB459301.1 | - | 3 - 4 | 384 | 116 |