The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316676.1 | NP_001303605.1 | 3 - 4 | 441 | 269 |
NM_006504.5 | NP_006495.1 | 2 - 3 | 270 | 98 | |
GenBank mRNA | AF406557.1 | - | 2 - 3 | 205 | 98 |
BC050062.1 | - | 3 - 4 | 349 | 269 | |
BI559814.1 | - | 2 - 3 | 277 | 98 |