The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021203.3 | NP_067026.3 | 6 - 7 | 839 | 105 |
GenBank mRNA | AF086914.1 | - | 5 - 6 | 577 | 105 |
AF141882.3 | - | 5 - 6 | 570 | 105 | |
AK027307.1 | - | 2 - 3 | 1082 | 105 | |
AK027525.1 | - | 5 - 6 | 570 | 105 | |
AK075531.1 | - | 5 - 6 | 608 | 105 | |
BC063001.1 | - | 5 - 6 | 582 | 105 | |
BC065299.1 | - | 5 - 6 | 554 | 105 |