Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC6
Entrez Gene ID
8030
Gene Name
coiled-coil domain containing 6
Gene Aliases
D10S170, H4, PTC, TPC, TST1
Chromosome Location
Chr.10: 59788747 - 59906656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005436.4 NP_005427.2 1 - 2 539 102
GenBank mRNA AK292593.1 - 1 - 2 102
BC022284.1 - 1 - 2 433 102
BC036757.2 - 1 - 2 102
BC064391.1 - 1 - 2 102
S72869.1 - 1 - 2 102

View Full Product Details