The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002039.3 | NP_002030.2 | 2 - 3 | 723 | 70 |
NM_207123.2 | NP_997006.1 | 2 - 3 | 723 | 70 | |
XM_006714167.3 | XP_006714230.1 | 2 - 3 | 1540 | 70 | |
XM_006714168.3 | XP_006714231.1 | 2 - 3 | 1204 | 70 | |
XM_017007966.1 | XP_016863455.1 | 3 - 4 | 7367 | 70 | |
XM_017007967.1 | XP_016863456.1 | 2 - 3 | 452 | 70 | |
XM_017007968.1 | XP_016863457.1 | 3 - 4 | 7365 | 70 | |
XM_017007969.1 | XP_016863458.1 | 2 - 3 | 2616 | 70 | |
XM_017007970.1 | XP_016863459.1 | 2 - 3 | 2371 | 70 | |
XM_017007971.1 | XP_016863460.1 | 2 - 3 | 2371 | 70 | |
GenBank mRNA | AK289767.1 | - | 2 - 3 | 666 | 70 |
AK295684.1 | - | 2 - 3 | 367 | 70 | |
AK314635.1 | - | 2 - 3 | 403 | 70 | |
BC064848.1 | - | 2 - 3 | 587 | 70 | |
U43885.1 | - | 2 - 3 | 485 | 70 |