Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN3
Entrez Gene ID
9369
Gene Name
neurexin 3
Gene Aliases
C14orf60
Chromosome Location
Chr.14: 77979904 - 79868290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105250.2 NP_001098720.1 4 - 5 1648 80
NR_073546.1 4 - 5 1648 80
NR_073547.1 17 - 18 4682 80
XM_005268218.2 XP_005268275.1 20 - 21 4751 80
XM_006720322.2 XP_006720385.1 19 - 20 4739 80
XM_006720323.2 XP_006720386.1 18 - 19 4715 80
XM_011537363.1 XP_011535665.1 20 - 21 4751 80
XM_011537364.2 XP_011535666.1 21 - 22 4713 80
XM_011537365.2 XP_011535667.1 20 - 21 4539 80
XM_011537366.1 XP_011535668.1 19 - 20 4727 80
XM_011537368.1 XP_011535670.1 20 - 21 4751 80
XM_011537369.1 XP_011535671.1 20 - 21 4751 80
XM_011537370.1 XP_011535672.1 19 - 20 4727 80
XM_011537371.1 XP_011535673.1 20 - 21 4751 80
XM_011537372.1 XP_011535674.1 20 - 21 4751 80
XM_011537377.2 XP_011535679.1 16 - 17 3043 80
XM_017021790.1 XP_016877279.1 21 - 22 4851 80
XM_017021791.1 XP_016877280.1 21 - 22 4634 80
XM_017021792.1 XP_016877281.1 19 - 20 4534 80
XM_017021793.1 XP_016877282.1 19 - 20 4724 80
XM_017021794.1 XP_016877283.1 18 - 19 4715 80
XM_017021795.1 XP_016877284.1 18 - 19 4510 80
XM_017021796.1 XP_016877285.1 18 - 19 4700 80
XM_017021797.1 XP_016877286.1 17 - 18 4691 80
XM_017021798.1 XP_016877287.1 17 - 18 4688 80
XM_017021803.1 XP_016877292.1 19 - 20 4730 80
XM_017021804.1 XP_016877293.1 19 - 20 4724 80
XM_017021805.1 XP_016877294.1 18 - 19 4715 80
XM_017021806.1 XP_016877295.1 18 - 19 4700 80
GenBank mRNA AJ316284.2 - 17 - 18 4470 80
AJ493127.1 - 4 - 5 724 80
BC142649.1 - 4 - 5 1081 80
BC150194.1 - 4 - 5 1081 80

View Full Product Details