The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 12 - 13 | 1349 | 89 |
NM_020320.4 | NP_064716.2 | 13 - 14 | 1184 | 89 | |
NR_134857.1 | 13 - 14 | 1180 | 89 | ||
XM_011535948.2 | XP_011534250.1 | 13 - 14 | 1169 | 89 | |
XM_011535949.2 | XP_011534251.1 | 13 - 14 | 1169 | 89 | |
XM_017011072.1 | XP_016866561.1 | 13 - 14 | 1350 | 89 | |
XM_017011073.1 | XP_016866562.1 | 14 - 15 | 1568 | 89 | |
XM_017011074.1 | XP_016866563.1 | 13 - 14 | 1171 | 89 | |
XM_017011075.1 | XP_016866564.1 | 12 - 13 | 1294 | 89 | |
XM_017011076.1 | XP_016866565.1 | 14 - 15 | 1512 | 89 | |
XM_017011077.1 | XP_016866566.1 | 12 - 13 | 1071 | 89 | |
XM_017011078.1 | XP_016866567.1 | 12 - 13 | 1115 | 89 | |
XM_017011079.1 | XP_016866568.1 | 13 - 14 | 1350 | 89 | |
XM_017011080.1 | XP_016866569.1 | 14 - 15 | 1512 | 89 | |
XM_017011081.1 | XP_016866570.1 | 12 - 13 | 1115 | 89 | |
GenBank mRNA | AK023550.1 | - | 13 - 14 | 1127 | 89 |
AK315669.1 | - | 13 - 14 | 1121 | 89 | |
BC010420.1 | - | 13 - 14 | 1155 | 89 | |
BC105784.1 | - | 12 - 13 | 1288 | 89 |