The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 15 - 16 | 1659 | 112 |
NM_020320.4 | NP_064716.2 | 16 - 17 | 1494 | 112 | |
NR_134857.1 | 16 - 17 | 1490 | 112 | ||
XM_011535948.2 | XP_011534250.1 | 16 - 17 | 1479 | 112 | |
XM_017011072.1 | XP_016866561.1 | 16 - 17 | 1660 | 112 | |
XM_017011073.1 | XP_016866562.1 | 17 - 18 | 1878 | 112 | |
XM_017011074.1 | XP_016866563.1 | 16 - 17 | 1481 | 112 | |
XM_017011075.1 | XP_016866564.1 | 15 - 16 | 1604 | 112 | |
XM_017011076.1 | XP_016866565.1 | 17 - 18 | 1822 | 112 | |
XM_017011077.1 | XP_016866566.1 | 15 - 16 | 1381 | 112 | |
XM_017011078.1 | XP_016866567.1 | 15 - 16 | 1425 | 112 | |
XM_017011079.1 | XP_016866568.1 | 16 - 17 | 1660 | 112 | |
XM_017011080.1 | XP_016866569.1 | 17 - 18 | 1822 | 112 | |
XM_017011081.1 | XP_016866570.1 | 15 - 16 | 1425 | 112 | |
GenBank mRNA | AK023550.1 | - | 16 - 17 | 1437 | 112 |
AK093934.1 | - | 16 - 17 | 1867 | 112 | |
AK315669.1 | - | 16 - 17 | 1431 | 112 | |
BC010420.1 | - | 16 - 17 | 1465 | 112 | |
BC105784.1 | - | 15 - 16 | 1598 | 112 | |
CA415020.1 | - | 4 - 5 | 112 |