The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318533.1 | NP_001305462.1 | 14 - 15 | 1371 | 70 |
NM_017868.3 | NP_060338.3 | 14 - 15 | 1353 | 70 | |
XM_005271604.3 | XP_005271661.1 | 14 - 15 | 1569 | 70 | |
XM_011542890.2 | XP_011541192.1 | 15 - 16 | 1567 | 70 | |
XM_017017943.1 | XP_016873432.1 | 14 - 15 | 1353 | 70 | |
XM_017017944.1 | XP_016873433.1 | 15 - 16 | 1923 | 70 | |
XM_017017945.1 | XP_016873434.1 | 15 - 16 | 1406 | 70 | |
XM_017017946.1 | XP_016873435.1 | 14 - 15 | 1576 | 70 | |
XM_017017947.1 | XP_016873436.1 | 14 - 15 | 1353 | 70 | |
XM_017017948.1 | XP_016873437.1 | 14 - 15 | 1353 | 70 | |
XM_017017949.1 | XP_016873438.1 | 14 - 15 | 1353 | 70 | |
XM_017017950.1 | XP_016873439.1 | 14 - 15 | 1353 | 70 | |
GenBank mRNA | AK000542.1 | - | 13 - 14 | 1245 | 70 |
AK023921.1 | - | 14 - 15 | 1299 | 70 | |
AK091887.1 | - | 16 - 17 | 1965 | 70 | |
AK223117.1 | - | 13 - 14 | 1245 | 70 | |
AK292331.1 | - | 14 - 15 | 1291 | 70 | |
BC011546.1 | - | 13 - 14 | 1295 | 70 | |
BC032355.1 | - | 13 - 14 | 1275 | 70 |