Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS26B
Entrez Gene ID
112936
Gene Name
VPS26, retromer complex component B
Gene Aliases
Pep8b
Chromosome Location
Chr.11: 134224605 - 134247792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052875.4 NP_443107.1 3 - 4 1062 97
GenBank mRNA AF452718.1 - 3 - 4 1000 97
BC009747.2 - 3 - 4 978 97
BC013255.1 - 2 - 3 263 97
BC014128.2 - 3 - 4 987 97
BC098386.1 - 3 - 4 1063 97

View Full Product Details