Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIG10L
Entrez Gene ID
147645
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
-
Chromosome Location
Chr.19: 51331541 - 51342124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163922.1 NP_001157394.1 7 - 8 2306 59
GenBank mRNA BC016993.1 - 4 - 5 924 59
BC032383.1 - 4 - 5 918 59
BC049212.1 - 4 - 5 971 59

View Full Product Details