Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA17
Entrez Gene ID
128153
Gene Name
spermatogenesis associated 17
Gene Aliases
IQCH, MSRG-11, MSRG11
Chromosome Location
Chr.1: 217631324 - 217868151 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138796.3 NP_620151.1 7 - 8 778 92
XM_005273052.1 XP_005273109.1 7 - 8 760 92
XM_006711165.3 XP_006711228.1 6 - 7 718 92
GenBank mRNA AK098591.1 - 7 - 8 758 92
AK310291.1 - 10 - 11 1100 92
AY963797.1 - 7 - 8 781 92
BC014608.2 - 7 - 8 749 92

View Full Product Details