Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr.10: 32446082 - 32882864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026383.2 NP_001021554.1 1 - 1 52 94
NM_001321115.1 NP_001308044.1 1 - 1 52 94
NM_145023.5 NP_659460.3 1 - 1 52 94
NR_109826.1 1 - 1 52 94
NR_109827.1 1 - 1 52 94
GenBank mRNA HY009015.1 - 1 - 1 53 94

View Full Product Details