The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177381.1 | NP_001170852.1 | 10 - 11 | 2796 | 67 |
NM_001177382.1 | NP_001170853.1 | 11 - 12 | 2877 | 67 | |
NM_001177383.1 | NP_001170854.1 | 10 - 11 | 2787 | 67 | |
NM_001177384.1 | NP_001170855.1 | 9 - 10 | 2772 | 67 | |
NM_182485.2 | NP_872291.2 | 10 - 11 | 2853 | 67 | |
NM_182646.2 | NP_872587.2 | 9 - 10 | 2763 | 67 | |
XM_005248135.3 | XP_005248192.2 | 11 - 12 | 3109 | 67 | |
XM_011513777.2 | XP_011512079.1 | 10 - 11 | 3085 | 67 | |
XM_017007733.1 | XP_016863222.1 | 11 - 12 | 1245 | 67 | |
XM_017007734.1 | XP_016863223.1 | 11 - 12 | 1246 | 67 | |
GenBank mRNA | AY247744.1 | - | 10 - 11 | 1629 | 67 |
AY255519.1 | - | 9 - 10 | 1539 | 67 | |
BC103939.1 | - | 11 - 12 | 1468 | 67 | |
BC103940.1 | - | 10 - 11 | 1591 | 67 | |
BC103941.1 | - | 10 - 11 | 1528 | 67 | |
BC103942.1 | - | 12 - 13 | 1549 | 67 | |
BC105925.1 | - | 9 - 10 | 1510 | 67 |