The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307941.1 | NP_001294870.1 | 3 - 4 | 434 | 123 |
NM_033280.3 | NP_150596.1 | 3 - 4 | 434 | 123 | |
XM_011526260.2 | XP_011524562.1 | 4 - 5 | 441 | 123 | |
XM_017026073.1 | XP_016881562.1 | 3 - 4 | 295 | 123 | |
GenBank mRNA | AF212233.1 | - | 3 - 4 | 399 | 123 |
AK295346.1 | - | 3 - 4 | 396 | 123 | |
AK314107.1 | - | 3 - 4 | 406 | 123 | |
BC009703.1 | - | 3 - 4 | 386 | 123 | |
BQ417397.1 | - | 3 - 4 | 327 | 123 |