The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020157.3 | NP_064542.1 | 1 - 2 | 162 | 139 |
XM_017027959.1 | XP_016883448.1 | 1 - 2 | 138 | 139 | |
GenBank mRNA | AF233261.1 | - | 1 - 2 | 157 | 139 |
AF243505.1 | - | 1 - 2 | 120 | 139 | |
AJ242552.1 | - | 1 - 2 | 113 | 139 | |
AY359082.1 | - | 1 - 2 | 150 | 139 | |
BC101688.1 | - | 1 - 2 | 162 | 139 | |
BC101690.1 | - | 1 - 2 | 162 | 139 |