The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018728.3 | NP_061198.2 | 22 - 23 | 3030 | 106 |
XM_011521781.2 | XP_011520083.1 | 22 - 23 | 2946 | 106 | |
XM_017022408.1 | XP_016877897.1 | 21 - 22 | 2895 | 106 | |
XM_017022409.1 | XP_016877898.1 | 17 - 18 | 2337 | 106 | |
XM_017022410.1 | XP_016877899.1 | 21 - 22 | 2880 | 106 | |
GenBank mRNA | AF272390.1 | - | 22 - 23 | 2887 | 106 |
AK304136.1 | - | 21 - 22 | 2832 | 106 | |
BC144611.1 | - | 21 - 22 | 2649 | 106 |