The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018728.3 | NP_061198.2 | 6 - 7 | 908 | 88 |
XM_011521781.2 | XP_011520083.1 | 6 - 7 | 908 | 88 | |
XM_017022408.1 | XP_016877897.1 | 6 - 7 | 908 | 88 | |
GenBank mRNA | AF272390.1 | - | 6 - 7 | 765 | 88 |
AK304136.1 | - | 7 - 8 | 1063 | 88 | |
BC064841.1 | - | 6 - 7 | 854 | 88 |