The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152718.2 | NP_689931.2 | 14 - 15 | 2062 | 81 |
XM_017017340.1 | XP_016872829.1 | 14 - 15 | 1999 | 81 | |
XM_017017341.1 | XP_016872830.1 | 13 - 14 | 1933 | 81 | |
XM_017017342.1 | XP_016872831.1 | 13 - 14 | 1655 | 81 | |
XM_017017343.1 | XP_016872832.1 | 15 - 16 | 1846 | 81 | |
GenBank mRNA | AK056571.1 | - | 14 - 15 | 1820 | 81 |
AK302241.1 | - | 3 - 4 | 301 | 81 | |
AY205235.1 | - | 12 - 13 | 1567 | 81 | |
BC142624.1 | - | 14 - 15 | 1809 | 81 |