The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005061.2 | NP_005052.1 | 5 - 6 | 735 | 84 |
XM_011522571.2 | XP_011520873.1 | 5 - 6 | 1195 | 84 | |
GenBank mRNA | AK313162.1 | - | 5 - 6 | 747 | 84 |
BC050413.1 | - | 5 - 6 | 727 | 84 | |
U65581.1 | - | 5 - 6 | 736 | 84 |