The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005061.2 | NP_005052.1 | 7 - 8 | 1000 | 68 |
XM_011522571.2 | XP_011520873.1 | 7 - 8 | 1460 | 68 | |
GenBank mRNA | AK313162.1 | - | 7 - 8 | 1012 | 68 |
BC050413.1 | - | 7 - 8 | 992 | 68 | |
U65581.1 | - | 7 - 8 | 1001 | 68 |