The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020390.5 | NP_065123.1 | 4 - 5 | 530 | 114 |
GenBank mRNA | AK311962.1 | - | 4 - 5 | 491 | 114 |
AY205258.1 | - | 4 - 5 | 530 | 114 | |
AY205259.1 | - | 4 - 5 | 530 | 114 | |
AY205260.1 | - | 4 - 5 | 530 | 114 | |
AY205261.1 | - | 4 - 5 | 530 | 114 | |
BC036072.1 | - | 4 - 5 | 501 | 114 |