The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 5 - 6 | 659 | 136 |
GenBank mRNA | AK297669.1 | - | 5 - 6 | 571 | 136 |
AK297674.1 | - | 5 - 6 | 512 | 136 | |
AK314817.1 | - | 5 - 6 | 653 | 136 | |
BC027881.1 | - | 5 - 6 | 645 | 136 | |
V01514.1 | - | 5 - 6 | 659 | 136 |