The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 6 - 7 | 751 | 131 |
GenBank mRNA | AK297669.1 | - | 6 - 7 | 663 | 131 |
AK297674.1 | - | 6 - 7 | 604 | 131 | |
AK314817.1 | - | 6 - 7 | 745 | 131 | |
BC027881.1 | - | 6 - 7 | 737 | 131 | |
V01514.1 | - | 6 - 7 | 751 | 131 |