Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
Chromosome Location
Chr.4: 73436219 - 73455785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134.2 NP_001125.1 9 - 10 1237 73
GenBank mRNA AK297669.1 - 9 - 10 1149 73
AK297674.1 - 9 - 10 1090 73
AK314817.1 - 9 - 10 1231 73
BC027881.1 - 9 - 10 1223 73
V01514.1 - 9 - 10 1237 73

View Full Product Details